Methanol
Methanol
What Is Methanol?
Methanol is the simplest alcohol: a methyl group linked to a hydroxyl group. Light, colorless, highly flammable, and volatile, it is commonly used in scientific research as a solvent, de-colorant, or denaturant.
Although methanol was used for embalming by ancient Egyptians, pure methanol was isolated by Robert Boyle in 1661 and its composition determined by Jean-Baptiste Dumas and Eugene Peligot in 1834.
Available in a variety of grades and percentages, methanol is also used as a reagent in the synthesis of chemicals such as acetic acid or formaldehyde.
Methanol is used:
  • In many analytical wet lab techniques, including chromatography, spectometry, and chemical synthesis 
  • To manufacture paints, adhesives, foams, and textiles 
  • For fuel and to make biodiesel 
  • As a denaturant for industrially produced ethanol 
Although similar to ethanol (drinking alcohol) in appearance and odor, methanol is toxic to the nervous system. Methanol that is ingested, inhaled, or otherwise absorbed can lead to blindness, coma, and death.
When working with methanol, the environment must be properly ventilated, and personal protective equipment (PPE) must be used to protect the eyes, face, and skin. Methanol containers should not be left open to the air and closed containers should be stored in dry, cool, and well-ventilated locations. To extinguish methanol fires, use dry chemical, carbon dioxide, water spray, or alcohol-resistant foam.
Synonyms: Methyl alcohol, wood alcohol, carbinol, wood spirit, 67-56-1, methylol, methyl hydroxide, MeOH, methyl hydrate
View All Methanol Products
solvents-methanol-0959
    Physical and Chemical Properties
    PropertyValue  
    Property
    Molecular Weight
    Value
    32.04 g/mol
    Property
    Molar Mass
    Value
    32.042 g/mol
    Property
    Density
    Value
    0.791 g/mL at 25ºC
    Property
    Boiling Point
    Value
    64.7ºC
    Property
    Melting Point
    Value
    -98ºC
    Property
    Flash Point
    Value
    11.1ºC
    Property
    Viscosity
    Value
    0.544 cP at 25ºC
    Property
    Dielectric Constant
    Value
    33.00 at 20ºC
    Property
    Dipole Moment
    Value
    1.7
    Property
    UV Cutoff
    Value
    210 nm
    Property
    Refractive Index
    Value
    1.329 at 20ºC
    Property
    Vapor Density
    Value
    1.11 (vs. air)
    Property
    Exact Mass
    Value
    32.026215 g/mol
    Property
    Monoisotopic Mass
    Value
    32.026215 g/mol
    Uses
    Methanol is highly versatile and a critical component of olefins, formaldehyde, and hundreds of other chemicals.
    About 40 percent of the methanol produced worldwide is used to make formaldehyde, which is ultimately used in plastics, plywood, paints, explosives, textiles, and many other products.
    Methanol is a traditional denaturant for ethanol and used as a solvent, antifreeze ingredient, and windshield washer fluid. It is also used in automotive and marine fuel, DME and bioDME, biodiesel, fuel cells, electricity, and boilers and cookstoves.
    Designations and Grades
    PropertyValue  
    Property
    CAS Number
    Value
    67-56-1
    Property
    MDL Number
    Value
    MFCD00004595
    Property
    RIDAD /UN Number
    Value
    UN 1230 3/PG 2
    Property
    Beilstein Registry
    Value
    1098229
    Property
    EG/EC Number
    Value
    2006596
    Property
    NSC Number
    Value
    85232
    Property
    ICSC Number
    Value
    0057
    Property
    RTECS Number
    Value
    85232
    Grades
    ACS, ACS Reagent, Certified ACS
    Anhydrous/Dry/Extra Dry
    Biochemical
    Electronic/Semiconductor
    GC, Capillary GC
    GC Gradient
    GC/Pesticide Residue
    Histological
    HPLC Gradient
    HPLC/UHPLC
    Laboratory/Technical
    LC-MS, Ultra LC-MS
    Low Water
    Mobile Phase
    NF/USP
    Purge and Trap
    Sequencing
    Spectrophotometry
    Solvent Blends
    Trace Metals
    A New Way to Shop for Chemicals
    Conduct your search by drawing your molecule or uploading its chemical structure.
    Search Now
    Chemical Safety at Your Fingertips
    Locate a safety data sheet (SDS) for a particular chemical or catalog number.
    Search Now