Hexane
What Is Hexane?
Hexane, also called hexanes, is a mixture of straight-chain alkanes with six carbon atoms. It consists of approximately 60 percent n-hexane, varying amounts of the isomers 2-methylpentane and 3-methylpentane, and smaller amounts of non-isomer alkanes. Hexane is colorless and odorless when pure and has a boiling point of roughly 69°C (156°F).
This solvent cannot be easily deprotonated and is therefore used in labs for reactions involving very strong bases, like preparing organolithiums. Hexane can also extract oil and grease from water and soil. In chromatography, hexane is used as a non-polar solvent, but its alkanes may interfere with analyses if the sample becomes contaminated.
Hexane is obtained primarily by crude oil refinement and its composition depends on the source of the oil and the refining process.
Physical and Chemical Properties
| Property | Value | ||
|---|---|---|---|
Molecular Weight | 86.18 g/mol | ||
Density | 0.6606 g/mL 25°C | ||
Boiling Point | 68.73°C | ||
Melting Point | -95.3°C | ||
Flash Point | -22°C | ||
Viscosity | 0.300 cP at 25ºC | ||
Dielectric Constant | 1.890 at 20°C | ||
UV Cutoff | 210 nm | ||
Refractive Index | 1.3727 at 25°C | ||
Vapor Pressure | 13 mm Hg at 20°C | ||
Water Solubility | 0.0095 mg/mL at 25°C | ||
Heat of Combustion | 4163.2 kJ/mol | ||
Heat of Vaporization | 1.56 kJ/mol at 25°C | ||
Surface Tension | 17.89 mN/m at 25°C |
How Is Hexane Used?
Hexane is used in textile manufacturing to make glues and adhesives for shoes, leather products, and roofing. It’s also used for cleansing and degreasing and to extract cooking oils, like canola or soy, from seeds.
Designations and Grades
| Property | Value | ||
|---|---|---|---|
CAS Number | 110-54-3 | ||
MDL Number | MFCD00009520 | ||
RIDAD/UN Number | 1208 | ||
Beilstein Registry | 1730733 | ||
EG/EC Number | 203-777-6 | ||
NSC Number | 8472 | ||
ICSC Number | 0279 | ||
RTECS Number | MN9275000 |
Grades
ACS/ACS Reagent/Certified ACS
Biochemical
Dry/Extra Dry/Molecular Sieve
Electronic
Environmental
GC/Pesticide Residue/HR-GC
HPLC/HPLC Gradient/LC-MS
Laboratory/Technical
Pesticide
Spectrophotometry
A New Way to Shop for Chemicals
Conduct your search by drawing your molecule or uploading its chemical structure.
Chemical Safety at Your Fingertips
Locate a safety data sheet (SDS) for a particular chemical or catalog number.