Uncover the Range of Novel Genetic Events in Your Sequence Data with Unrivaled Simplicity
Since their inception, genetic analyzers have evolved to support diverse applications that extend beyond long-read DNA sequencing and fragment analysis. Whether you are looking for a single mutation, investigating a new genome, or profiling gene expression, capillary electrophoresis (CE) genetic analyzers can be customized for your diverse research endeavors.
SeqStudio: Rapidly Access an Array of Applications
The latest innovation for Sanger sequencing and fragment analysis is the Thermo Scientific SeqStudio Genetic Analyzer. It features a click-in cartridge with four capillaries that comes pre-loaded with a universal polymer, making it easier and faster to perform Sanger sequencing and fragment analysis in the same run. Plus, SeqStudio offers an intuitive software interface on a benchtop platform that minimizes hands-on time and enables ready access to various research applications.
Plasmid Sequencing
- Accurately analyze subcloned inserts
- Get flexible read lengths ranging from <300 bp to >600 bp
Oncology
- Profile cancers and detect low level variants
- Discover low-level variants at frequencies as low as 5% with the sensitivity and accuracy of Sanger sequencing
Genome Editing
- Reliably detect genome editing or CRISPR-mediated events
- Analyze the efficiency of events using the tracking of Indels by decomposition (TIDE) software
Next-Generation Sequencing (NGS) Confirmation
- Rapidly confirm the variants identified by NGS
- Import .vcf files
- Verify variants using the cloud-based NGC module
Human Cell Line Authentication
- Bypass errors due to contamination
- Integrate with Identifiler Plus and Identifiler Direct workflows to safeguard research quality
Multiplex Ligation-Dependent Probe Amplification (MLPA)
- Determine copy number across multiple DNA sequences
Species Identification
- Identify species in an unknown sample by sequencing DNA of “fingerprint”
- Use the 16S ribosomal RNA (rRNA) gene, now considered to be the gold-standard method for bacteria taxonomic classification and identification
The new SeqStudio Genetic Analyzer offers access to these applications in an easy-to-use platform that streamlines your CE experiments and minimizes hands-on time. The cloud-based sharing expands your impact and helps increase your efficiency.