UserName
Thermo Scientific Logo
Thermo Scientific CO2 Incubators
Culture Confidently
Thermo Scientific CO2 incubators let you culture with confidence — day after day and year after year. They offer innovative features like solutions to support your GMP and cleanroom needs, HEPA air filtration that surrounds cells with clean room-like air quality, and a choice of 100% pure copper or polished stainless-steel interior surfaces. Plus, the direct-heat units feature a high-temperature decontamination function to eliminate the need to handle or autoclave components.
    Shop Thermo Scientific CO2 Incubators
    Cleanroom & Specialty CO2 Incubators
    Designed for Compatibility and Control
    Choose from CO2 incubators designed to support your cleanroom and other specialty needs, including GMP compatibility, high-volume culturing, and high contamination control.
    Heracell VIOS 160i/250i CR CO2 Incubators
    Tri-Gas CO2 Incubators
    Variable Oxygen Control
    Many cell cultures thrive best in CO2 incubators with controlled levels of oxygen. Select an O2 option to simulate physiological hypoxic environments (for stem cell and IVF applications) or choose to increase oxygen concentration for the ability to operate at hyperoxic levels.
    Direct Heat CO2 Incubators
    Even Temperatures
    Direct heat CO2 incubators feature efficient, high-performance heaters located on every chamber surface, providing even temperature distribution throughout the entire chamber.
    Forma Steri-Cult CO2 Incubators
    Water-Jacketed CO2 Incubators
    Dual Insulation
    Water-jacketed CO2 incubators offer unique triple-wall construction, providing temperature stability with dual layers of water and insulation.
    Resources
    Filename
    heracell-vios-cr-co2-incubator-design-smart-note.pdf
    Size
    759 KB
    Format
    application/pdf
    Filename
    cts-series-co2-incubators-cleaning-disinfection-application-note.pdf
    Size
    517 KB
    Format
    application/pdf
    Filename
    thermo-scientific-co2-incubators-humidity-water-smart-note.pdf
    Size
    649 KB
    Format
    application/pdf
    Filename
    thermo-scientific-copper-incubators-brochure.pdf
    Size
    711 KB
    Format
    application/pdf
    Filename
    thermo-scientific-steri-run-sterilization-application-notes.pdf
    Size
    2 MB
    Format
    application/pdf
    Filename
    thermo-scientific-incubator-care-instruction.pdf
    Size
    1 MB
    Format
    application/pdf
    Filename
    thermo-co2-incubator-circulating-fan-flyer.pdf
    Size
    204 KB
    Format
    application/pdf
    Filename
    thermo-co2-incubator-variable-o2-brochure.pdf
    Size
    1012 KB
    Format
    application/pdf
    Filename
    thermo-scientific-co2-incubators-decontamination-smart-note.pdf
    Size
    317 KB
    Format
    application/pdf
    Videos
    Thermo Scientific CO2 Incubators | Fisher Scientific
    Thermo Scientific CO2 Incubators | Fisher Scientific
    Thermo Scientific CO2 Incubators | Fisher Scientific
    Thermo Scientific CO2 Incubators | Fisher Scientific