accessibility menu, dialog, popup

UserName

CH3H3CCH3BrBr(E/Z)

CAS RN 99584-95-9

IUPAC Name: 1,2-dibromo-3,3-dimethylbut-1-ene
Synonyms: 1,2-dibromo-3,3-dimethylbut-1-ene
Molecular Weight (g/mol): 241.95
Molecular Formula: C6H10Br2
InChi Key: JDGBPINHTJNHMH-UHFFFAOYSA-N
SMILES: CC(C)(C)C(Br)=CBr

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results