accessibility menu, dialog, popup

UserName

CAS RN 99455-05-7

IUPAC Name: 6-bromo-2-methoxyquinoline
Synonyms: 6-bromo-2-methoxyquinoline
Molecular Weight (g/mol): 238.08
Molecular Formula: C10H8BrNO
InChi Key: KBTKKEMYFUMFSJ-UHFFFAOYSA-N
SMILES: COC1=CC=C2C=C(Br)C=CC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results