CAS RN 987-78-0
IUPAC Name:
{2-[({[(2R,3S,4R,5R)-5-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl phosphonato)oxy]ethyl}trimethylazanium
Synonyms:
citicoline cdp-colina citicolinum citicolina citidin difosfato de colina 胞磷胆碱 cytidindiphosphocholin cdp-choline
Molecular Weight (g/mol):
488.33
Molecular Formula:
C14H26N4O11P2
InChi Key:
RZZPDXZPRHQOCG-OJAKKHQRSA-N
SMILES:
C[N+](C)(C)CCOP([O-])(=O)OP(O)(=O)OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=CC(N)=NC1=O
1
–
1
of
1
results
1
Promotions Available
Citicoline, 98%
Citicoline, 98%
CAS: 987-78-0 Molecular Formula: C14H26N4O11P2 Molecular Weight (g/mol): 488.33 MDL Number: MFCD00868097 InChI Key: RZZPDXZPRHQOCG-OJAKKHQRSA-N Synonym: cdp-choline,cidifos,citicoline,cytidine diphosphate choline,cdp-choline 1-,5'-o-2-trimethylazaniumyl ethoxy phosphinato oxy phosphinato cytidine PubChem CID: 25202509 ChEBI: CHEBI:58779 IUPAC Name: [[(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] 2-(trimethylazaniumyl)ethyl phosphate SMILES: C[N+](C)(C)CCOP([O-])(=O)OP(O)(=O)OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=CC(N)=NC1=O