accessibility menu, dialog, popup

UserName

CAS RN 98026-88-1

IUPAC Name: 2-chloropyrimidine-5-sulfonyl chloride
Synonyms: 2-chloropyrimidine-5-sulfonyl chloride
Molecular Weight (g/mol): 213.03
Molecular Formula: C4H2Cl2N2O2S
InChi Key: DMAOAJYJIATWJS-UHFFFAOYSA-N
SMILES: ClC1=NC=C(C=N1)S(Cl)(=O)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)