accessibility menu, dialog, popup

UserName

ClSiClCl

CAS RN 98-12-4

IUPAC Name: trichloro(cyclohexyl)silane
Synonyms: silane, trichlorocyclohexyl-
Molecular Weight (g/mol): 217.59
Molecular Formula: C6H11Cl3Si
InChi Key: SIPHWXREAZVVNS-UHFFFAOYSA-N
SMILES: Cl[Si](Cl)(Cl)C1CCCCC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results