accessibility menu, dialog, popup

UserName

CAS RN 97858-62-3

CH3OPPOH3C

CAS RN 97858-62-3

IUPAC Name: (2-methoxyphenyl)({2-[(2-methoxyphenyl)(phenyl)phosphanyl]ethyl})phenylphosphane
Synonyms: dipamp
Molecular Weight (g/mol): 458.48
Molecular Formula: C28H28O2P2
InChi Key: QKZWXPLBVCKXNQ-UHFFFAOYNA-N
SMILES: COC1=CC=CC=C1P(CCP(C1=CC=CC=C1)C1=CC=CC=C1OC)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results