accessibility menu, dialog, popup

UserName

CH3OOFFFFFF

CAS RN 96617-71-9

IUPAC Name: ethyl 3,5-bis(trifluoromethyl)benzoate
Synonyms: ethyl 3,5-bis(trifluoromethyl)benzoate
Molecular Weight (g/mol): 286.17
Molecular Formula: C11H8F6O2
InChi Key: NBWZJMOEFTYMOF-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)