CAS RN 96386-92-4
IUPAC Name:
(3S)-3-amino-5-methylhexanoic acid hydrochloride
Synonyms:
(3S)-3-amino-5-methylhexanoic acid hydrochloride
Molecular Weight (g/mol):
181.66
Molecular Formula:
C7H16ClNO2
InChi Key:
NXVYPYHWONGEFQ-RGMNGODLSA-N
SMILES:
Cl.CC(C)C[C@H](N)CC(O)=O
1
–
1
of
1
results
1
Promotions Available
L-beta-Homoleucine hydrochloride, 95%
L-beta-Homoleucine hydrochloride, 95%
CAS: 96386-92-4 Molecular Formula: C7H16ClNO2 Molecular Weight (g/mol): 181.66 MDL Number: MFCD01862871 InChI Key: NXVYPYHWONGEFQ-RGMNGODLSA-N Synonym: l-beta-homoleucine hydrochloride,s-3-amino-5-methylhexanoic acid hydrochloride,h-beta-holeu-oh.hcl,3s-3-amino-5-methylhexanoic acid hydrochloride,h-?-holeu-oh.hcl,hexanoic acid, 3-amino-5-methyl-, hydrochloride, 3s,l-beta-homoleucine-hcl,l-beta-homoleucinehcl,h-b-holeu-oh hcl,h-,a-holeu-oh.hcl PubChem CID: 2761524 IUPAC Name: (3S)-3-amino-5-methylhexanoic acid hydrochloride SMILES: Cl.CC(C)C[C@H](N)CC(O)=O