CAS RN 959996-58-8
IUPAC Name:
6-(trifluoromethyl)pyridine-3-sulfonyl chloride
Synonyms:
6-(trifluoromethyl)pyridine-3-sulfonyl chloride
Molecular Weight (g/mol):
245.6
Molecular Formula:
C6H3ClF3NO2S
InChi Key:
NSGLNIQAWVNZFB-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=NC=C(C=C1)S(Cl)(=O)=O
1
–
1
of
1
results
1
Promotions Available
6-(Trifluoromethyl)pyridine-3-sulfonyl chloride, 97%
6-(Trifluoromethyl)pyridine-3-sulfonyl chloride, 97%
CAS: 959996-58-8 Molecular Formula: C6H3ClF3NO2S Molecular Weight (g/mol): 245.6 MDL Number: MFCD10699112 InChI Key: NSGLNIQAWVNZFB-UHFFFAOYSA-N Synonym: 6-trifluoromethyl pyridine-3-sulfonyl chloride,6-trifluoromethyl-3-pyridinesulfonyl chloride,3-pyridinesulfonylchloride,6-trifluoromethyl,chloro 6-trifluoromethyl 3-pyridyl sulfone,3-pyridinesulfonyl chloride, 6-trifluoromethyl PubChem CID: 45594311 IUPAC Name: 6-(trifluoromethyl)pyridine-3-sulfonyl chloride SMILES: C1=CC(=NC=C1S(=O)(=O)Cl)C(F)(F)F