accessibility menu, dialog, popup

UserName

H3CPCH3H3COH3CFFFSOONSOOFFF

CAS RN 959698-44-3

IUPAC Name: tributyl(2-methoxyethyl)phosphanium; trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
Synonyms: bistriflylimide anion; tributyl(2-methoxyethyl)phosphanium
Molecular Weight (g/mol): 541.55
Molecular Formula: C17H34F6NO5PS2
InChi Key: QFLRMCUVYQFPCO-UHFFFAOYSA-N
SMILES: FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.CCCC[P+](CCCC)(CCCC)CCOC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results