CAS RN 957060-90-1
IUPAC Name:
[2-(2,2,2-trifluoroethoxy)phenyl]boronic acid
Synonyms:
2-(2,2,2-trifluoroethoxy)phenylboronic acid
Molecular Weight (g/mol):
219.95
Molecular Formula:
C8H8BF3O3
InChi Key:
IFWDRBWGMUCFMH-UHFFFAOYSA-N
SMILES:
OB(O)C1=CC=CC=C1OCC(F)(F)F
1
–
1
of
1
results
1
Promotions Available
2-(2,2,2-Trifluoroethoxy)benzeneboronic acid, 98%, Thermo Scientific Chemicals
2-(2,2,2-Trifluoroethoxy)benzeneboronic acid, 98%, Thermo Scientific Chemicals
CAS: 957060-90-1 Molecular Formula: C8H8BF3O3 Molecular Weight (g/mol): 219.954 MDL Number: MFCD09027258 InChI Key: IFWDRBWGMUCFMH-UHFFFAOYSA-N Synonym: 2-2,2,2-trifluoroethoxy phenylboronic acid,2-2,2,2-trifluoroethoxy benzeneboronic acid,2-2,2,2-trifluoroethoxy phenyl boronic acid,acmc-209s1a,2-2,2,2-trifluoroethoxy phenyl boronicacid PubChem CID: 43145273 IUPAC Name: [2-(2,2,2-trifluoroethoxy)phenyl]boronic acid SMILES: B(C1=CC=CC=C1OCC(F)(F)F)(O)O