accessibility menu, dialog, popup

UserName

CAS RN 955-83-9

O

CAS RN 955-83-9

IUPAC Name: 2,5-diphenylfuran
Synonyms: 2,5-diphenylfuran PPF
Molecular Weight (g/mol): 220.27
Molecular Formula: C16H12O
InChi Key: VUPDHIIPAKIKAB-UHFFFAOYSA-N
SMILES: O1C(=CC=C1C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results