accessibility menu, dialog, popup

UserName

CAS RN 95349-35-2

H3CSiOOH

CAS RN 95349-35-2

IUPAC Name: 2-[benzyl(methyl)phenylsilyl]acetic acid
Synonyms: [benzyl(methyl)phenylsilyl]acetic acid
Molecular Weight (g/mol): 270.4
Molecular Formula: C16H18O2Si
InChi Key: JIPQZCPYHVQEIZ-UHFFFAOYNA-N
SMILES: C[Si](CC(O)=O)(CC1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results