accessibility menu, dialog, popup

UserName

CAS RN 952616-39-6

HNOOOOH

CAS RN 952616-39-6

IUPAC Name: 3-{[(benzyloxy)carbonyl]amino}cyclohexane-1-carboxylic acid
Synonyms: 3-{[(benzyloxy)carbonyl]amino}cyclohexane-1-carboxylic acid
Molecular Weight (g/mol): 277.32
Molecular Formula: C15H19NO4
InChi Key: LTEORMGXUMWZCU-UHFFFAOYNA-N
SMILES: OC(=O)C1CCCC(C1)NC(=O)OCC1=CC=CC=C1
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results