CAS RN 95166-77-1
IUPAC Name:
1-(2-phenylethenyl)-4-(2-phenylethyl)benzene
Synonyms:
1-(2-phenylethenyl)-4-(2-phenylethyl)benzene
Molecular Weight (g/mol):
284.4
Molecular Formula:
C22H20
InChi Key:
VKIHNQGTZXZCPE-UHFFFAOYSA-N
SMILES:
C(CC1=CC=C(C=CC2=CC=CC=C2)C=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
1-(trans-2-Phenylethenyl)-4-(2-phenylethyl)benzene, 97%
1-(trans-2-Phenylethenyl)-4-(2-phenylethyl)benzene, 97%
CAS: 95166-77-1 Molecular Formula: C22H20 Molecular Weight (g/mol): 284.402 MDL Number: MFCD12407080 InChI Key: VKIHNQGTZXZCPE-ACCUITESSA-N Synonym: 1-trans-2-phenylethenyl-4-2-phenylethyl benzene,1-e-2-phenylethenyl-4-2-phenylethyl benzene,4-phenethyl-cis-stilbene,4-phenethyl-trans-stilbene,4-trans-beta-styryl bibenzyl PubChem CID: 59832893 IUPAC Name: 1-[(E)-2-phenylethenyl]-4-(2-phenylethyl)benzene SMILES: C1=CC=C(C=C1)CCC2=CC=C(C=C2)C=CC3=CC=CC=C3