CAS RN 950603-35-7
IUPAC Name:
1-[5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl]piperidine-4-carboxylic acid
Synonyms:
1-[5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl]piperidine-4-carboxylic acid
Molecular Weight (g/mol):
281.25
Molecular Formula:
C9H10F3N3O2S
InChi Key:
PWQMNLMMMCREOV-UHFFFAOYSA-N
SMILES:
OC(=O)C1CCN(CC1)C1=NN=C(S1)C(F)(F)F
1
–
2
of
2
results
2
1-[5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl]piperidine-4-carboxylic acid, Thermo Scientific™
CAS: 950603-35-7 Molecular Formula: C9H10F3N3O2S Molecular Weight (g/mol): 281.25 MDL Number: MFCD11101440 InChI Key: PWQMNLMMMCREOV-UHFFFAOYSA-N Synonym: 1-5-trifluoromethyl-1,3,4-thiadiazol-2-yl piperidine-4-carboxylic acid PubChem CID: 33589561 IUPAC Name: 1-[5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl]piperidine-4-carboxylic acid SMILES: OC(=O)C1CCN(CC1)C1=NN=C(S1)C(F)(F)F