accessibility menu, dialog, popup

UserName

CAS RN 950603-17-5

H3CHNO

CAS RN 950603-17-5

IUPAC Name: [(3,4-dihydro-2H-1-benzopyran-6-yl)methyl](methyl)amine
Synonyms: (3,4-dihydro-2H-1-benzopyran-6-ylmethyl)(methyl)amine
Molecular Weight (g/mol): 177.25
Molecular Formula: C11H15NO
InChi Key: OYZVOCNCDRSGDZ-UHFFFAOYSA-N
SMILES: CNCC1=CC2=C(OCCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)