accessibility menu, dialog, popup

UserName

CH3NCH3ClN

CAS RN 95-89-6

IUPAC Name: 3-chloro-2,5-dimethylpyrazine
Synonyms: 3-chloro-2,5-dimethylpyrazine
Molecular Weight (g/mol): 142.59
Molecular Formula: C6H7ClN2
InChi Key: NNBALVIZMGWZHS-UHFFFAOYSA-N
SMILES: CC1=CN=C(C)C(Cl)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results