accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CNNH2FFF

CAS RN 947249-01-6

IUPAC Name: 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3-(trifluoromethyl)pyridin-2-amine
Synonyms: 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3-(trifluoromethyl)pyridin-2-amine
Molecular Weight (g/mol): 288.08
Molecular Formula: C12H16BF3N2O2
InChi Key: OQZKROYNCVLJKM-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(=C(N)N=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results