accessibility menu, dialog, popup

UserName

CAS RN 946409-35-4

IUPAC Name: tert-butyl 4-{[4-(aminomethyl)phenyl]methoxy}piperidine-1-carboxylate
Synonyms: tert-butyl 4-{[4-(aminomethyl)phenyl]methoxy}piperidine-1-carboxylate
Molecular Weight (g/mol): 320.43
Molecular Formula: C18H28N2O3
InChi Key: NZWAUTDCSSMVGG-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCC(CC1)OCC1=CC=C(CN)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)