accessibility menu, dialog, popup

UserName

CH3H3CCH3OOHNNBr

CAS RN 943722-24-5

IUPAC Name: tert-butyl N-[(5-bromopyridin-3-yl)methyl]carbamate
Synonyms: tert-butyl N-[(5-bromopyridin-3-yl)methyl]carbamate
Molecular Weight (g/mol): 287.16
Molecular Formula: C11H15BrN2O2
InChi Key: VZPOHASZICQBDX-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCC1=CC(Br)=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)