CAS RN 941716-90-1
IUPAC Name:
methyl({[4-(phenoxymethyl)phenyl]methyl})amine
Synonyms:
methyl({[4-(phenoxymethyl)phenyl]methyl})amine
Molecular Weight (g/mol):
227.31
Molecular Formula:
C15H17NO
InChi Key:
PSMWUOSOWNRPOJ-UHFFFAOYSA-N
SMILES:
CNCC1=CC=C(COC2=CC=CC=C2)C=C1
1
–
1
of
1
results
1
N-Methyl-4-(phenoxymethyl)benzylamine, 95%, Thermo Scientific™
CAS: 941716-90-1 Molecular Formula: C15H17NO Molecular Weight (g/mol): 227.31 MDL Number: MFCD09879942 InChI Key: PSMWUOSOWNRPOJ-UHFFFAOYSA-N Synonym: n-methyl-4-phenoxymethyl benzylamine,methyl 4-phenoxymethyl phenyl methyl amine,n-methyl-1-4-phenoxymethyl phenyl methanamine PubChem CID: 24229704 IUPAC Name: methyl({[4-(phenoxymethyl)phenyl]methyl})amine SMILES: CNCC1=CC=C(COC2=CC=CC=C2)C=C1