accessibility menu, dialog, popup

UserName

CH3O

CAS RN 94-65-5

IUPAC Name: 2-propylcyclohexan-1-one
Synonyms: cyclohexanone, 2-propyl-
Molecular Weight (g/mol): 140.23
Molecular Formula: C9H16O
InChi Key: OCJLPZCBZSCVCO-UHFFFAOYNA-N
SMILES: CCCC1CCCCC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results