CAS RN 937796-11-7
IUPAC Name:
[2-(pyrrolidin-1-yl)pyrimidin-5-yl]methanol
Synonyms:
[2-(pyrrolidin-1-yl)pyrimidin-5-yl]methanol
Molecular Weight (g/mol):
179.22
Molecular Formula:
C9H13N3O
InChi Key:
SJXXWSAQNABDEB-UHFFFAOYSA-N
SMILES:
OCC1=CN=C(N=C1)N1CCCC1
1
–
2
of
2
results
2
(2-Pyrrolidin-1-ylpyrimidin-5-yl)methanol, 97%, Thermo Scientific™
CAS: 937796-11-7 Molecular Formula: C9H13N3O Molecular Weight (g/mol): 179.223 MDL Number: MFCD09863240 InChI Key: SJXXWSAQNABDEB-UHFFFAOYSA-N Synonym: 2-pyrrolidin-1-ylpyrimidin-5-yl methanol,2-pyrrolidin-1-yl pyrimidin-5-yl methanol,5-hydroxymethyl-2-pyrrolidin-1-yl pyrimidine,2-pyrrolidinylpyrimidin-5-yl methan-1-ol,5-hydroxymethyl-2-pyrrolidin-1-ylpyrimidine PubChem CID: 24229767 IUPAC Name: (2-pyrrolidin-1-ylpyrimidin-5-yl)methanol SMILES: C1CCN(C1)C2=NC=C(C=N2)CO