CAS RN 937-52-0
IUPAC Name:
4-phenylbutan-2-amine
Synonyms:
α-benzylisopropylamine benzenepropanamine, �-methyl-
Molecular Weight (g/mol):
149.24
Molecular Formula:
C10H15N
InChi Key:
WECUIGDEWBNQJJ-UHFFFAOYNA-N
SMILES:
CC(N)CCC1=CC=CC=C1
1
–
1
of
1
results
1
(R)-(-)-1-Methyl-3-phenylpropylamine, 98%
CAS: 937-52-0 Molecular Formula: C10H15N Molecular Weight (g/mol): 149.24 MDL Number: MFCD00145208 InChI Key: WECUIGDEWBNQJJ-UHFFFAOYNA-N Synonym: r---1-methyl-3-phenylpropylamine,r-4-phenylbutan-2-amine,2r-4-phenylbutan-2-amine,unii-o4f6h42uwq,o4f6h42uwq,r-1-methyl-3-phenylpropylamine,r-4-phenylbutane-2-amine,4-phenylbutan-2-amine, r,r-1-methyl-3-phenyl-propylamine PubChem CID: 2734033 SMILES: CC(N)CCC1=CC=CC=C1