accessibility menu, dialog, popup

UserName

CAS RN 934570-52-2

CH3H3CCH3OONOOHO

CAS RN 934570-52-2

IUPAC Name: 4-[({1-[(tert-butoxy)carbonyl]piperidin-4-yl}oxy)methyl]benzoic acid
Synonyms: 4-({[1-(tert-butoxycarbonyl)piperidin-4-yl]oxy}methyl)benzoic acid
Molecular Weight (g/mol): 335.4
Molecular Formula: C18H25NO5
InChi Key: CGLNCSVUPUKDKT-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCC(CC1)OCC1=CC=C(C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)