accessibility menu, dialog, popup

UserName

H3CNCH3OHOH

CAS RN 93413-62-8

IUPAC Name: 4-[2-(dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]phenol
Synonyms: desvenlafaxine pristiq
Molecular Weight (g/mol): 263.38
Molecular Formula: C16H25NO2
InChi Key: KYYIDSXMWOZKMP-UHFFFAOYNA-N
SMILES: CN(C)CC(C1=CC=C(O)C=C1)C1(O)CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results