accessibility menu, dialog, popup

UserName

CAS RN 934-37-2

IUPAC Name: 2-methylimidazo[1,2-a]pyridine
Synonyms: 2-methylimidazo[1,2-a]pyridine
Molecular Weight (g/mol): 132.17
Molecular Formula: C8H8N2
InChi Key: BZACBBRLMWHCNM-UHFFFAOYSA-N
SMILES: CC1=CN2C=CC=CC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results