accessibility menu, dialog, popup

UserName

CAS RN 934-31-6

IUPAC Name: 2-oxo-2H-1,3-dithiole-4,5-dicarbonitrile
Synonyms: 2-oxo-1,3-dithiole-4,5-dicarbonitrile
Molecular Weight (g/mol): 168.19
Molecular Formula: C5N2OS2
InChi Key: BPDMXHRQYNWQSI-UHFFFAOYSA-N
SMILES: O=C1SC(C#N)=C(S1)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results