CAS RN 933-76-6
IUPAC Name:
4,5-dichloro-2-methyl-2,3-dihydropyridazin-3-one
Synonyms:
4,5-dichloro-2-methylpyridazin-3-one
Molecular Weight (g/mol):
179
Molecular Formula:
C5H4Cl2N2O
InChi Key:
ACKBTCUMGAHRIE-UHFFFAOYSA-N
SMILES:
CN1N=CC(Cl)=C(Cl)C1=O
1
–
1
of
1
results
1
Promotions Available
4,5-Dichloro-2-methyl-3(2H)-pyridazinone, 97%, Thermo Scientific Chemicals
4,5-Dichloro-2-methyl-3(2H)-pyridazinone, 97%, Thermo Scientific Chemicals
CAS: 933-76-6 Molecular Formula: C5H4Cl2N2O Molecular Weight (g/mol): 179.00 MDL Number: MFCD00051686 InChI Key: ACKBTCUMGAHRIE-UHFFFAOYSA-N Synonym: 4,5-dichloro-2-methylpyridazin-3 2h-one,4,5-dichloro-2-methyl-3 2h-pyridazinone,3 2h-pyridazinone, 4,5-dichloro-2-methyl,4,5-dichloro-2-methyl-2h-pyridazin-3-one,4,5-dichloro-2-methyl-2,3-dihydropyridazin-3-one,3 2h-pyridazinone,4,5-dichloro-2-methyl,4,5-dichloro-2-methyl-2-hydropyridazin-3-one,pubchem16243,acmc-209wiz,5-24-02-00023 beilstein handbook reference PubChem CID: 120462 IUPAC Name: 4,5-dichloro-2-methylpyridazin-3-one SMILES: CN1N=CC(Cl)=C(Cl)C1=O