accessibility menu, dialog, popup

UserName

SOO

CAS RN 932-41-2

IUPAC Name: thiophene-2,3-dicarbaldehyde
Synonyms: thiophene-2,3-dicarbaldehyde
Molecular Weight (g/mol): 140.16
Molecular Formula: C6H4O2S
InChi Key: WSEJZRIZDQWMKQ-UHFFFAOYSA-N
SMILES: O=CC1=C(C=O)C=CS1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results