accessibility menu, dialog, popup

UserName

CAS RN 926921-78-0

IUPAC Name: 5-(chloromethyl)-2-(thiophen-2-yl)pyrimidine
Synonyms: 5-(chloromethyl)-2-(thiophen-2-yl)pyrimidine
Molecular Weight (g/mol): 210.68
Molecular Formula: C9H7ClN2S
InChi Key: HBGUDWOIHDHRCQ-UHFFFAOYSA-N
SMILES: ClCC1=CN=C(N=C1)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results