accessibility menu, dialog, popup

UserName

NOHOHO

CAS RN 923169-37-3

IUPAC Name: 4-(hydroxymethyl)pyridine-2-carboxylic acid
Synonyms: 4-(hydroxymethyl)pyridine-2-carboxylic acid
Molecular Weight (g/mol): 153.14
Molecular Formula: C7H7NO3
InChi Key: QMLYYPGJXCVEHA-UHFFFAOYSA-N
SMILES: OCC1=CC(=NC=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results