accessibility menu, dialog, popup

UserName

CAS RN 922-28-1

H3CCH3CH3CH3

CAS RN 922-28-1

IUPAC Name: 3,4-dimethylheptane
Synonyms: heptane, 3,4-dimethyl-
Molecular Weight (g/mol): 128.26
Molecular Formula: C9H20
InChi Key: MAKRYGRRIKSDES-UHFFFAOYNA-N
SMILES: CCCC(C)C(C)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results