accessibility menu, dialog, popup

UserName

CAS RN 92-46-6

H3CClN

CAS RN 92-46-6

IUPAC Name: 6-chloro-2-methylquinoline
Synonyms: 6-chloro-2-methylquinoline
Molecular Weight (g/mol): 177.63
Molecular Formula: C10H8ClN
InChi Key: OCCIBGIEIBQGAJ-UHFFFAOYSA-N
SMILES: CC1=CC=C2C=C(Cl)C=CC2=N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results