accessibility menu, dialog, popup

UserName

CH3NNOH

CAS RN 910037-09-1

IUPAC Name: [3-(1-methyl-1H-pyrazol-3-yl)phenyl]methanol
Synonyms: [3-(1-methylpyrazol-3-yl)phenyl]methanol
Molecular Weight (g/mol): 188.23
Molecular Formula: C11H12N2O
InChi Key: IFFXGZCJGGVFOF-UHFFFAOYSA-N
SMILES: CN1C=CC(=N1)C1=CC(CO)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)