accessibility menu, dialog, popup

UserName

CH3HNONCH3CH3

CAS RN 910037-03-5

IUPAC Name: dimethyl(3-{3-[(methylamino)methyl]phenoxy}propyl)amine
Synonyms: dimethyl(3-{3-[(methylamino)methyl]phenoxy}propyl)amine
Molecular Weight (g/mol): 222.33
Molecular Formula: C13H22N2O
InChi Key: ODWHICGCLWFLBT-UHFFFAOYSA-N
SMILES: CNCC1=CC(OCCCN(C)C)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)