accessibility menu, dialog, popup

UserName

N

CAS RN 91-73-6

IUPAC Name: N,N-dibenzylaniline
Synonyms: N,N-dibenzylaniline
Molecular Weight (g/mol): 273.38
Molecular Formula: C20H19N
InChi Key: ISGXOWLMGOPVPB-UHFFFAOYSA-N
SMILES: C(N(CC1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)