accessibility menu, dialog, popup

UserName

CH3NCH3HO

CAS RN 91-68-9

IUPAC Name: 3-(diethylamino)phenol
Synonyms: phenol, 3-(diethylamino)-
Molecular Weight (g/mol): 165.24
Molecular Formula: C10H15NO
InChi Key: WAVOOWVINKGEHS-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC=CC(O)=C1
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)