accessibility menu, dialog, popup

UserName

OOHS

CAS RN 906352-94-1

IUPAC Name: [4-(thiophen-2-yl)oxan-4-yl]methanol
Synonyms: [4-(thiophen-2-yl)oxan-4-yl]methanol
Molecular Weight (g/mol): 198.28
Molecular Formula: C10H14O2S
InChi Key: JHVRUEHWCGLJPP-UHFFFAOYSA-N
SMILES: OCC1(CCOCC1)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)