CAS RN 906352-60-1
CAS RN 906352-60-1
IUPAC Name:
2-[3-(chloromethyl)phenyl]-1,3-thiazole
Synonyms:
2-[3-(chloromethyl)phenyl]-1,3-thiazole
Molecular Weight (g/mol):
209.69
Molecular Formula:
C10H8ClNS
InChi Key:
SWICJACRAPFUKX-UHFFFAOYSA-N
SMILES:
ClCC1=CC=CC(=C1)C1=NC=CS1
1
–
1
of
1
results
1
2-[3-(Chloromethyl)phenyl]-1,3-thiazole, 97%, Thermo Scientific™
CAS: 906352-60-1 Molecular Formula: C10H8ClNS Molecular Weight (g/mol): 209.69 MDL Number: MFCD09025829 InChI Key: SWICJACRAPFUKX-UHFFFAOYSA-N Synonym: 2-3-chloromethyl phenyl-1,3-thiazole,2-3-chloromethyl phenyl thiazole PubChem CID: 18525728 IUPAC Name: 2-[3-(chloromethyl)phenyl]-1,3-thiazole SMILES: ClCC1=CC=CC(=C1)C1=NC=CS1