CAS RN 90600-20-7
IUPAC Name:
(2S)-2-{[(tert-butoxy)carbonyl]amino}pent-4-enoate
Synonyms:
(2S)-2-[(tert-butoxycarbonyl)amino]pent-4-enoate
Molecular Weight (g/mol):
214.24
Molecular Formula:
C10H16NO4
InChi Key:
BUPDPLXLAKNJMI-ZETCQYMHSA-M
SMILES:
CC(C)(C)OC(=O)N[C@@H](CC=C)C([O-])=O
1
–
1
of
1
results
1
Promotions Available
(S)-N-BOC-Allylglycine, 95%, Thermo Scientific Chemicals
(S)-N-BOC-Allylglycine, 95%, Thermo Scientific Chemicals
CAS: 90600-20-7 Molecular Formula: C10H17NO4 Molecular Weight (g/mol): 215.249 MDL Number: MFCD01320851 InChI Key: BUPDPLXLAKNJMI-ZETCQYMHSA-N Synonym: s-2-tert-butoxycarbonyl amino pent-4-enoic acid,s-n-boc-allylglycine,boc-l-allylglycine,boc-alpha-allyl-l-gly,4-pentenoic acid, 2-1,1-dimethylethoxy carbonyl amino-, 2s,s-2-tert-butoxycarbonylamino pent-4-enoic acid,2s-2-tert-butoxycarbonyl amino pent-4-enoic acid,boc-l-alpha-allyl-gly,boc--allyl-l-gly,rarechem bk pt 0250 PubChem CID: 2734487 IUPAC Name: (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pent-4-enoic acid SMILES: CC(C)(C)OC(=O)NC(CC=C)C(=O)O