accessibility menu, dialog, popup

UserName

CAS RN 90555-66-1

BOCH3HOOH

CAS RN 90555-66-1

IUPAC Name: (3-ethoxyphenyl)boronic acid
Synonyms: 3-ethoxyphenylboronic acid
Molecular Weight (g/mol): 165.98
Molecular Formula: C8H11BO3
InChi Key: CHCWUTJYLUBETR-UHFFFAOYSA-N
SMILES: CCOC1=CC=CC(=C1)B(O)O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results