accessibility menu, dialog, popup

UserName

A(a)

CAS RN 9046-10-0

IUPAC Name: pyrrolidine-3,4-diol
Synonyms: pyrrolidine-3,4-diol
Molecular Weight (g/mol): 103.12
Molecular Formula: C4H9NO2
InChi Key: JCZPOYAMKJFOLA-UHFFFAOYNA-N
SMILES: OC1CNCC1O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)