CAS RN 9008-42-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Promotions Available
Thermo Scientific Chemicals Silver Protein for Histology, Strong
Thermo Scientific Chemicals Silver Protein for Histology, Strong
CAS: 9008-42-8 Synonym: Protargol