CAS RN 90-42-6
CAS RN 90-42-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Cyclohexylcyclohexanone 97.0+%, TCI America™
CAS: 90-42-6 Molecular Formula: C12H20O Molecular Weight (g/mol): 180.29 MDL Number: MFCD00013795 InChI Key: UOBQDYFTAJKQAL-UHFFFAOYNA-N PubChem CID: 7016 IUPAC Name: [1,1'-bi(cyclohexane)]-2-one SMILES: O=C1CCCCC1C1CCCCC1